Assay Details
Gene Symbol: anapc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016392.2 | NP_001016392.1 | 3 - 4 | 261 | 69 | |
XM_012962247.2 | XP_012817701.1 | 4 - 5 | 464 | 69 | ||
XM_012962250.2 | XP_012817704.1 | 4 - 5 | 345 | 69 | ||
XM_012962253.1 | XP_012817707.1 | 4 - 5 | 371 | 69 | ||
GenBank mRNA | BC160495.1 | - | 4 - 5 | 371 | 69 | |
BC170586.1 | - | 3 - 4 | 246 | 69 | ||
BC170941.1 | - | 3 - 4 | 246 | 69 | ||
CR761172.2 | - | 3 - 4 | 261 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
