Assay Details
| Gene Symbol: c1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016424.2 | NP_001016424.1 | 2 - 3 | 178 | 80 | |
| GenBank mRNA | CR761067.2 | - | 2 - 3 | 178 | 80 | |
More Information
Set Membership: |
|

c1d
Genomic Map