Assay Details
Gene Symbol: manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016425.2 | NP_001016425.1 | 1 - 2 | 109 | 65 | |
GenBank mRNA | BC170885.1 | - | 1 - 2 | 109 | 65 | |
BC170889.1 | - | 1 - 2 | 109 | 65 | ||
CR761066.2 | - | 1 - 2 | 109 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
