Assay Details
Gene Symbol: rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016437.2 | NP_001016437.1 | 1 - 2 | 221 | 109 | |
GenBank mRNA | BC170899.1 | - | 1 - 2 | 152 | 109 | |
BC170903.1 | - | 1 - 2 | 152 | 109 | ||
CR761033.2 | - | 1 - 2 | 221 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
