Assay Details
| Gene Symbol: mpzl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016455.2 | NP_001016455.1 | 5 - 6 | 650 | 93 | |
| GenBank mRNA | BC159352.1 | - | 5 - 6 | 666 | 93 | |
| CR760972.2 | - | 5 - 6 | 650 | 93 | ||
More Information
Set Membership: |
|

mpzl2
Genomic Map