Assay Details
| Gene Symbol: rab5if | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016462.2 | NP_001016462.1 | 1 - 2 | 134 | 81 | |
| GenBank mRNA | BC091613.1 | - | 1 - 2 | 156 | 81 | |
| CR760947.2 | - | 1 - 2 | 134 | 81 | ||
More Information
Set Membership: |
|

rab5if
Genomic Map