Assay Details
Gene Symbol: tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016476.2 | NP_001016476.1 | 3 - 4 | 851 | 71 | |
XM_012962377.2 | XP_012817831.1 | 4 - 5 | 1047 | 71 | ||
GenBank mRNA | BC135907.1 | - | 3 - 4 | 781 | 71 | |
CR760901.2 | - | 3 - 4 | 851 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
