Assay Details
Gene Symbol: bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016502.2 | NP_001016502.1 | 2 - 3 | 299 | 92 | |
GenBank mRNA | BC135374.1 | - | 2 - 3 | 290 | 92 | |
CR760796.2 | - | 2 - 3 | 299 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
