Assay Details
Gene Symbol: gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016512.2 | NP_001016512.1 | 4 - 5 | 607 | 66 | |
XM_012961097.2 | XP_012816551.1 | 5 - 6 | 1533 | 66 | ||
XM_012961098.2 | XP_012816552.1 | 5 - 6 | 1531 | 66 | ||
XM_012961099.2 | XP_012816553.1 | 6 - 7 | 1607 | 66 | ||
XM_012961100.2 | XP_012816554.1 | 5 - 6 | 1527 | 66 | ||
XM_012961102.2 | XP_012816556.1 | 5 - 6 | 1524 | 66 | ||
XM_012961103.2 | XP_012816557.1 | 5 - 6 | 1532 | 66 | ||
XM_012961104.2 | XP_012816558.1 | 5 - 6 | 1523 | 66 | ||
XM_018093050.1 | XP_017948539.1 | 4 - 5 | 730 | 66 | ||
GenBank mRNA | BC170710.1 | - | 3 - 4 | 560 | 66 | |
BC170712.1 | - | 3 - 4 | 560 | 66 | ||
CR926227.2 | - | 4 - 5 | 607 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
