Assay Details
Gene Symbol: rhbdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016521.2 | NP_001016521.1 | 867 | 75 | ||
GenBank mRNA | BC170807.1 | - | 7 - 8 | 801 | 75 | |
BC170809.1 | - | 7 - 8 | 801 | 75 | ||
CR926207.2 | - | 867 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
