Assay Details
Gene Symbol: cotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016541.2 | NP_001016541.1 | 3 - 4 | 419 | 85 | |
GenBank mRNA | BC135722.1 | - | 3 - 4 | 481 | 85 | |
CR760762.2 | - | 3 - 4 | 419 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
