Assay Details
| Gene Symbol: cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016543.2 | NP_001016543.1 | 1 - 2 | 109 | 75 | |
| GenBank mRNA | BC161502.1 | - | 1 - 2 | 161 | 75 | |
| CR760755.2 | - | 1 - 2 | 109 | 75 | ||
More Information
Set Membership: |
|

cox6a1
Genomic Map