Assay Details
| Gene Symbol: pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016545.2 | NP_001016545.1 | 9 - 10 | 1133 | 64 | |
| XM_012968421.1 | XP_012823875.1 | 9 - 10 | 1090 | 64 | ||
| XM_012968422.1 | XP_012823876.1 | 9 - 10 | 1221 | 64 | ||
| GenBank mRNA | CR760747.2 | - | 9 - 10 | 1133 | 64 | |
More Information
Set Membership: |
|

pgk1
Genomic Map