Assay Details
Gene Symbol: gs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016546.2 | NP_001016546.1 | 1 - 2 | 379 | 106 | |
GenBank mRNA | BC170623.1 | - | 1 - 2 | 377 | 106 | |
CR760742.2 | - | 1 - 2 | 379 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
