Assay Details
| Gene Symbol: ift57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016561.2 | NP_001016561.1 | 8 - 9 | 1005 | 72 | |
| XM_012956983.2 | XP_012812437.1 | 9 - 10 | 1109 | 72 | ||
| XM_012956984.2 | XP_012812438.1 | 9 - 10 | 1075 | 72 | ||
| XM_018091264.1 | XP_017946753.1 | 9 - 10 | 1280 | 72 | ||
| GenBank mRNA | BC170691.1 | - | 8 - 9 | 948 | 72 | |
| BC170695.1 | - | 8 - 9 | 948 | 72 | ||
| CR760693.2 | - | 8 - 9 | 1005 | 72 | ||
More Information
Set Membership: |
|

ift57
Genomic Map