Assay Details
Gene Symbol: c16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016564.2 | NP_001016564.1 | 2 - 3 | 351 | 62 | |
XM_018097108.1 | XP_017952597.1 | 2 - 3 | 342 | 62 | ||
GenBank mRNA | BC154871.1 | - | 1 - 2 | 369 | 62 | |
CR760682.2 | - | 2 - 3 | 351 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
