Assay Details
Gene Symbol: c16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016564.2 | NP_001016564.1 | 4 - 5 | 703 | 67 | |
XM_018097108.1 | XP_017952597.1 | 4 - 5 | 694 | 67 | ||
GenBank mRNA | BC154871.1 | - | 3 - 4 | 721 | 67 | |
CR760682.2 | - | 4 - 5 | 703 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
