Assay Details
| Gene Symbol: setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016577.2 | NP_001016577.1 | 10 - 11 | 1251 | 69 | |
| XM_012968426.2 | XP_012823880.1 | 11 - 12 | 1368 | 69 | ||
| GenBank mRNA | BC135194.1 | - | 7 - 8 | 867 | 69 | |
| BC171209.1 | - | 10 - 11 | 1163 | 69 | ||
| CR760640.2 | - | 10 - 11 | 1251 | 69 | ||
More Information
Set Membership: |
|

setd3
Genomic Map