Assay Details
| Gene Symbol: cnot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016578.2 | NP_001016578.1 | 2 - 3 | 296 | 68 | |
| GenBank mRNA | BC170730.1 | - | 1 - 2 | 222 | 68 | |
| BC170734.1 | - | 1 - 2 | 222 | 68 | ||
| CR760639.2 | - | 2 - 3 | 296 | 68 | ||
More Information
Set Membership: |
|

cnot8
Genomic Map