Assay Details
| Gene Symbol: psat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016582.2 | NP_001016582.1 | 8 - 9 | 1041 | 74 | |
| XM_012962521.2 | XP_012817975.1 | 8 - 9 | 1128 | 74 | ||
| GenBank mRNA | BC127350.1 | - | 8 - 9 | 1105 | 74 | |
| CR760626.2 | - | 8 - 9 | 1041 | 74 | ||
More Information
Set Membership: |
|

psat1
Genomic Map