Assay Details
| Gene Symbol: glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016597.2 | NP_001016597.1 | 2 - 3 | 349 | 74 | |
| GenBank mRNA | CR760576.2 | - | 2 - 3 | 349 | 74 | |
More Information
Set Membership: |
|

glrx
Genomic Map