Assay Details
| Gene Symbol: cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016617.2 | NP_001016617.1 | 1 - 2 | 129 | 66 | |
| GenBank mRNA | BC118689.1 | - | 1 - 2 | 93 | 66 | |
| CR760508.2 | - | 1 - 2 | 129 | 66 | ||
More Information
Set Membership: |
|

cbx6
Genomic Map