Assay Details
| Gene Symbol: uqcrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016654.2 | NP_001016654.1 | 1 - 2 | 173 | 85 | |
| XM_012959036.2 | XP_012814490.1 | 2 - 3 | 261 | 85 | ||
| GenBank mRNA | BC155017.1 | - | 2 - 3 | 258 | 85 | |
| BC155034.1 | - | 2 - 3 | 258 | 85 | ||
| BC170793.1 | - | 1 - 2 | 173 | 85 | ||
| BC170891.1 | - | 1 - 2 | 173 | 85 | ||
| CR760339.2 | - | 1 - 2 | 173 | 85 | ||
More Information
Set Membership: |
|

uqcrq
Genomic Map