Assay Details
Gene Symbol: hoxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016678.2 | NP_001016678.1 | 1 - 2 | 625 | 78 | |
GenBank mRNA | BC160395.1 | - | 1 - 2 | 634 | 78 | |
BC170961.1 | - | 1 - 2 | 609 | 78 | ||
BC170965.1 | - | 1 - 2 | 609 | 78 | ||
CR760220.2 | - | 1 - 2 | 625 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
