Assay Details
| Gene Symbol: rpap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016680.2 | NP_001016680.1 | 16 - 17 | 1936 | 73 | |
| XM_012959037.2 | XP_012814491.1 | 16 - 17 | 1993 | 73 | ||
| GenBank mRNA | BC121516.1 | - | 15 - 16 | 1915 | 73 | |
| CR760208.2 | - | 16 - 17 | 1936 | 73 | ||
More Information
Set Membership: |
|

rpap3
Genomic Map