Assay Details
| Gene Symbol: abo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016683.2 | NP_001016683.1 | 6 - 7 | 615 | 79 | |
| XM_012968437.2 | XP_012823891.1 | 6 - 7 | 680 | 79 | ||
| XM_012968438.2 | XP_012823892.1 | 7 - 8 | 598 | 79 | ||
| GenBank mRNA | BC168620.1 | - | 6 - 7 | 610 | 79 | |
| BC170657.1 | - | 5 - 6 | 313 | 79 | ||
| BC170661.1 | - | 5 - 6 | 313 | 79 | ||
| CR760204.2 | - | 6 - 7 | 615 | 79 | ||
More Information
Set Membership: |
|

abo
Genomic Map