Assay Details
| Gene Symbol: tuba4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016694.2 | NP_001016694.1 | 3 - 4 | 557 | 85 | |
| XM_012970434.2 | XP_012825888.1 | 3 - 4 | 534 | 85 | ||
| GenBank mRNA | BC170862.1 | - | 3 - 4 | 442 | 85 | |
| BC171015.1 | - | 3 - 4 | 442 | 85 | ||
| CR760170.2 | - | 3 - 4 | 557 | 85 | ||
More Information
Set Membership: |
|

tuba4b
Genomic Map