Assay Details
Gene Symbol: get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016708.2 | NP_001016708.1 | 1 - 2 | 190 | 73 | |
GenBank mRNA | BC135489.1 | - | 1 - 2 | 149 | 73 | |
CR760128.2 | - | 1 - 2 | 190 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
