Assay Details
Gene Symbol: cat.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016716.2 | NP_001016716.1 | 489 | 106 | ||
GenBank mRNA | CR760094.2 | - | 489 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
