Assay Details
| Gene Symbol: slc39a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016721.2 | NP_001016721.1 | 1 - 2 | 131 | 73 | |
| XM_012966622.2 | XP_012822076.1 | 2 - 3 | 215 | 73 | ||
| XM_018097746.1 | XP_017953235.1 | 2 - 3 | 215 | 73 | ||
| GenBank mRNA | CR760068.2 | - | 1 - 2 | 131 | 73 | |
More Information
Set Membership: |
|

slc39a11
Genomic Map