Assay Details
| Gene Symbol: rrad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016726.2 | NP_001016726.1 | 4 - 5 | 771 | 84 | |
| GenBank mRNA | BC135196.1 | - | 4 - 5 | 728 | 84 | |
| CR760056.2 | - | 4 - 5 | 771 | 84 | ||
More Information
Set Membership: |
|

rrad
Genomic Map