Assay Details
| Gene Symbol: mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016727.2 | NP_001016727.1 | 2 - 3 | 260 | 78 | |
| GenBank mRNA | BC158935.1 | - | 2 - 3 | 471 | 78 | |
| CR760046.2 | - | 2 - 3 | 260 | 78 | ||
More Information
Set Membership: |
|

mrpl43
Genomic Map