Assay Details
| Gene Symbol: rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016768.2 | NP_001016768.1 | 3 - 4 | 488 | 56 | |
| XM_012961123.2 | XP_012816577.1 | 3 - 4 | 523 | 56 | ||
| XM_012961124.1 | XP_012816578.1 | 3 - 4 | 547 | 56 | ||
| XM_012961125.2 | XP_012816579.1 | 3 - 4 | 492 | 56 | ||
| GenBank mRNA | BC158145.1 | - | 3 - 4 | 553 | 56 | |
| CR855743.2 | - | 3 - 4 | 488 | 56 | ||
More Information
Set Membership: |
|

rcn1
Genomic Map