Assay Details
| Gene Symbol: nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016772.2 | NP_001016772.1 | 5 - 6 | 667 | 59 | |
| XM_012966629.2 | XP_012822083.1 | 5 - 6 | 739 | 59 | ||
| XM_012966630.2 | XP_012822084.1 | 5 - 6 | 739 | 59 | ||
| XM_012966632.2 | XP_012822086.1 | 4 - 5 | 583 | 59 | ||
| XM_012966633.2 | XP_012822087.1 | 4 - 5 | 586 | 59 | ||
| XM_012966634.2 | XP_012822088.1 | 5 - 6 | 600 | 59 | ||
| XM_012966635.2 | XP_012822089.1 | 5 - 6 | 564 | 59 | ||
| XM_018095396.1 | XP_017950885.1 | 5 - 6 | 573 | 59 | ||
| GenBank mRNA | CR855733.2 | - | 5 - 6 | 667 | 59 | |
More Information
Set Membership: |
|

nmnat1
Genomic Map