Assay Details
| Gene Symbol: efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016778.2 | NP_001016778.1 | 3 - 4 | 548 | 75 | |
| XM_012964728.1 | XP_012820182.1 | 3 - 4 | 554 | 75 | ||
| GenBank mRNA | BC158143.1 | - | 3 - 4 | 475 | 75 | |
| CR855706.2 | - | 3 - 4 | 548 | 75 | ||
More Information
Set Membership: |
|

efcab1
Genomic Map