Assay Details
| Gene Symbol: washc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016824.2 | NP_001016824.1 | 1 - 2 | 124 | 90 | |
| GenBank mRNA | CR855531.2 | - | 1 - 2 | 124 | 90 | |
More Information
Set Membership: |
|

washc3
Genomic Map