Assay Details
Gene Symbol: washc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016824.2 | NP_001016824.1 | 6 - 7 | 588 | 92 | |
GenBank mRNA | CR855531.2 | - | 6 - 7 | 588 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
