Assay Details
Gene Symbol: mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016828.3 | NP_001016828.2 | 6 - 7 | 889 | 65 | |
GenBank mRNA | BC093462.1 | - | 6 - 7 | 889 | 65 | |
CR855516.2 | - | 6 - 7 | 908 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
