Assay Details
Gene Symbol: fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016838.2 | NP_001016838.1 | 9 - 10 | 805 | 68 | |
GenBank mRNA | BC121555.1 | - | 9 - 10 | 822 | 68 | |
CR855480.2 | - | 9 - 10 | 805 | 68 | ||
HQ171200.1 | - | 7 - 8 | 720 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
