Assay Details
| Gene Symbol: washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016854.2 | NP_001016854.1 | 4 - 5 | 605 | 66 | |
| GenBank mRNA | BC123023.1 | - | 4 - 5 | 573 | 66 | |
| CR855419.2 | - | 4 - 5 | 605 | 66 | ||
More Information
Set Membership: |
|

washc1
Genomic Map