Assay Details
| Gene Symbol: washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016854.2 | NP_001016854.1 | 5 - 6 | 739 | 84 | |
| XM_012959046.2 | XP_012814500.1 | 6 - 7 | 839 | 84 | ||
| XM_012959047.1 | XP_012814501.1 | 6 - 7 | 753 | 84 | ||
| XM_012959048.2 | XP_012814502.1 | 6 - 7 | 839 | 84 | ||
| GenBank mRNA | BC090140.1 | - | 1 - 2 | 57 | 84 | |
| BC123023.1 | - | 5 - 6 | 707 | 84 | ||
| CR855419.2 | - | 5 - 6 | 739 | 84 | ||
More Information
Set Membership: |
|

washc1
Genomic Map