Assay Details
| Gene Symbol: rap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016898.2 | NP_001016898.1 | 2 - 3 | 861 | 93 | |
| XM_012968445.2 | XP_012823899.1 | 3 - 4 | 1044 | 93 | ||
| XM_012968446.2 | XP_012823900.1 | 3 - 4 | 1042 | 93 | ||
| GenBank mRNA | CR848539.2 | - | 2 - 3 | 861 | 93 | |
More Information
Set Membership: |
|

rap2c
Genomic Map