Assay Details
| Gene Symbol: ccnq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012968449.2 | XP_012823903.1 | 3 - 4 | 385 | 104 | |
| XM_012968450.2 | XP_012823904.1 | 3 - 4 | 395 | 104 | ||
| XM_018096265.1 | XP_017951754.1 | 3 - 4 | 415 | 104 | ||
| GenBank mRNA | CR848270.2 | - | 3 - 4 | 472 | 104 | |
More Information
Set Membership: |
|

ccnq
Genomic Map