Assay Details
Gene Symbol: ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016919.2 | NP_001016919.1 | 1 - 2 | 210 | 75 | |
GenBank mRNA | BC161467.1 | - | 1 - 2 | 395 | 75 | |
CR848188.2 | - | 1 - 2 | 210 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
