Assay Details
Gene Symbol: spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016934.2 | NP_001016934.1 | 4 - 5 | 489 | 68 | |
XM_012957010.2 | XP_012812464.1 | 5 - 6 | 1145 | 68 | ||
XM_012957011.2 | XP_012812465.1 | 4 - 5 | 495 | 68 | ||
GenBank mRNA | CR762113.2 | - | 4 - 5 | 489 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
