Assay Details
Gene Symbol: ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016957.2 | NP_001016957.1 | 2 - 3 | 283 | 83 | |
GenBank mRNA | BC135315.1 | - | 2 - 3 | 313 | 83 | |
CR761461.2 | - | 2 - 3 | 283 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
