Assay Details
Gene Symbol: rfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016959.2 | NP_001016959.1 | 3 - 4 | 598 | 74 | |
GenBank mRNA | CR761430.2 | - | 3 - 4 | 598 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
