Assay Details
| Gene Symbol: znrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016977.2 | NP_001016977.1 | 3 - 4 | 586 | 65 | |
| GenBank mRNA | CR848382.2 | - | 3 - 4 | 586 | 65 | |
More Information
Set Membership: |
|

znrf1
Genomic Map