Assay Details
Gene Symbol: arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016978.2 | NP_001016978.1 | 3 - 4 | 480 | 67 | |
GenBank mRNA | BC135629.1 | - | 3 - 4 | 524 | 67 | |
CR848379.2 | - | 3 - 4 | 480 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
