Assay Details
Gene Symbol: cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016983.2 | NP_001016983.1 | 19 - 20 | 2376 | 66 | |
XM_012966644.1 | XP_012822098.1 | 20 - 21 | 2618 | 66 | ||
XM_012966645.1 | XP_012822099.1 | 20 - 21 | 2615 | 66 | ||
XM_012966646.1 | XP_012822100.1 | 20 - 21 | 2612 | 66 | ||
XM_012966647.2 | XP_012822101.1 | 20 - 21 | 2529 | 66 | ||
XM_012966649.1 | XP_012822103.1 | 19 - 20 | 2495 | 66 | ||
XM_012966650.1 | XP_012822104.1 | 19 - 20 | 2492 | 66 | ||
XM_012966651.1 | XP_012822105.1 | 19 - 20 | 2489 | 66 | ||
XM_012966652.2 | XP_012822106.1 | 19 - 20 | 2409 | 66 | ||
XM_012966653.2 | XP_012822107.1 | 19 - 20 | 2406 | 66 | ||
XM_012966654.2 | XP_012822108.1 | 19 - 20 | 2403 | 66 | ||
XM_012966655.2 | XP_012822109.1 | 19 - 20 | 2383 | 66 | ||
GenBank mRNA | CR848321.2 | - | 19 - 20 | 2376 | 66 | |
CR942612.2 | - | 16 - 17 | 1957 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
